Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCCTGGTGCTTCGGCTTCCTGG[C/T]GCTGGGCTACTTGCTCTACCTGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601745 | ||||||||||||||||||||
Literature Links: |
KCNK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNK1 - potassium two pore domain channel subfamily K member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002245.3 | 248 | Missense Mutation | GCG,GTG | A,V 27 | NP_002236.1 | |
XM_011544184.2 | 248 | Intron | XP_011542486.1 |
MIR4427 - microRNA 4427 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |