Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTAGTGCCATACAGTGAGGGCA[C/G]TGTCCTTTCCTAACCCAGTCTACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTAFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTAFR - platelet activating factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000952.4 | 2085 | UTR 3 | NP_000943.1 | |||
NM_001164721.1 | 2085 | UTR 3 | NP_001158193.1 | |||
NM_001164722.2 | 2085 | UTR 3 | NP_001158194.1 | |||
NM_001164723.2 | 2085 | UTR 3 | NP_001158195.1 |