Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTTAGGAACTAAAAAAAGAACT[A/G]TTCAAGTCTAGGATATTAAAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114230 MIM: 602143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN2 - calpain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TP53BP2 - tumor protein p53 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031685.2 | Intron | NP_001026855.2 | ||||
NM_005426.2 | Intron | NP_005417.1 | ||||
XM_011544268.1 | Intron | XP_011542570.1 | ||||
XM_011544269.2 | Intron | XP_011542571.1 | ||||
XM_017002220.1 | Intron | XP_016857709.1 |