Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATGTGCAGCAGAGGAAGGAGC[C/T]GCCCAGCACGGCCAGGCCAGCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610036 | ||||||||||||||||||||
Literature Links: |
CLDN19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN19 - claudin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123395.1 | 726 | Missense Mutation | AGC,GGC | S,G 179 | NP_001116867.1 | |
NM_001185117.1 | 726 | Silent Mutation | GCA,GCG | A,A 150 | NP_001172046.1 | |
NM_148960.2 | 726 | Missense Mutation | AGC,GGC | S,G 179 | NP_683763.2 |