Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGCAGCAGCAGCAACAGCAC[C/T]ACCCCGAGGACACCCCCGATCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159440 MIM: 602413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPZ - myelin protein zero | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000530.7 | 568 | Silent Mutation | GTA,GTG | V,V 168 | NP_000521.2 | |
NM_001315491.1 | 568 | Silent Mutation | GTA,GTG | V,V 168 | NP_001302420.1 | |
XM_017001321.1 | 568 | Silent Mutation | GTA,GTG | V,V 178 | XP_016856810.1 |
SDHC - succinate dehydrogenase complex subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |