Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATCTTGAAAAGGAGCTCTAGT[A/G]TTGTGGGCAATGACCCGCTGGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
AUNIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AUNIP - aurora kinase A and ninein interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287490.1 | 1004 | Silent Mutation | AAC,AAT | N,N 301 | NP_001274419.1 | |
NM_024037.2 | 1004 | Silent Mutation | AAC,AAT | N,N 301 | NP_076942.1 | |
XM_017002300.1 | 1004 | Silent Mutation | AAC,AAT | N,N 266 | XP_016857789.1 |
MTFR1L - mitochondrial fission regulator 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |