Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCAACTGGGCTCAGAATCAGTG[C/T]TCCATTTTTGAATTTTCCAACCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610584 | ||||||||||||||||||||
Literature Links: |
C1orf131 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf131 - chromosome 1 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300830.1 | 869 | Missense Mutation | ACA,GCA | T,A 274 | NP_001287759.1 | |
NM_152379.3 | 869 | Missense Mutation | ACA,GCA | T,A 275 | NP_689592.2 |
TRIM67 - tripartite motif containing 67 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |