Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCGGAGGGCAGCCGAGGTCACA[C/T]TGGGGGCGCTGGCGCCATATGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603418 MIM: 614760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR7A2 - aldo-keto reductase family 7 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320979.1 | 786 | Missense Mutation | NP_001307908.1 | |||
NM_003689.3 | 786 | Missense Mutation | NP_003680.2 |
PQLC2 - PQ loop repeat containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |