Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGGGCCCTGCAGCTCATCTTC[A/G]TCACATGCCCCTCGCTGCTGGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121012 MIM: 603324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJA4 - gap junction protein alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GJB3 - gap junction protein beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005752.1 | 865 | Missense Mutation | ATC,GTC | I,V 84 | NP_001005752.1 | |
NM_024009.2 | 865 | Missense Mutation | ATC,GTC | I,V 84 | NP_076872.1 |