Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGGGCAACGGGGAGGTGAAGA[C/T]GGGGGAGCTGGAGTGGCTCATGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610171 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALML6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALML6 - calmodulin like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005244729.3 | 1760 | Missense Mutation | ACG,ATG | T,M 77 | XP_005244786.2 | |
XM_011540841.2 | 1760 | Missense Mutation | ACG,ATG | T,M 38 | XP_011539143.1 |
CFAP74 - cilia and flagella associated protein 74 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM52 - transmembrane protein 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |