Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTCTCTTCCCAGCATTGGCGCT[C/T]TCATAGGCCTGTCCGTAGCAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM159A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM159A - family with sequence similarity 159 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042693.2 | 1613 | Missense Mutation | CTC,TTC | L,F 65 | NP_001036158.1 | |
XM_011541377.2 | 1613 | Missense Mutation | CTC,TTC | L,F 69 | XP_011539679.1 | |
XM_011541378.2 | 1613 | Missense Mutation | CTC,TTC | L,F 69 | XP_011539680.1 |