Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTATCAAGTGCACAAACGTGGT[T/G]TGGTGGAGTATGAAGCCCAGTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POLR3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR3C - RNA polymerase III subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303456.1 | 535 | Missense Mutation | NP_001290385.1 | |||
NM_006468.7 | 535 | Missense Mutation | NP_006459.3 | |||
XM_005272927.3 | 535 | Intron | XP_005272984.1 | |||
XM_005272928.3 | 535 | Intron | XP_005272985.1 | |||
XM_011509078.2 | 535 | Missense Mutation | XP_011507380.1 |
RNF115 - ring finger protein 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |