Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAAGTGTTCCTGATGTCCCAGA[C/T]GCGCACCGAGTTGTCCATGGAGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TAF5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF5L - TATA-box binding protein associated factor 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025247.1 | 1738 | Intron | NP_001020418.1 | |||
NM_014409.3 | 1738 | Missense Mutation | ATC,GTC | I,V 536 | NP_055224.1 | |
XM_005273099.4 | 1738 | Missense Mutation | ATC,GTC | I,V 536 | XP_005273156.1 | |
XM_005273100.2 | 1738 | Missense Mutation | ATC,GTC | I,V 444 | XP_005273157.1 | |
XM_011544162.1 | 1738 | Missense Mutation | ATC,GTC | I,V 465 | XP_011542464.1 | |
XM_011544163.1 | 1738 | Missense Mutation | ATC,GTC | I,V 444 | XP_011542465.1 | |
XM_011544164.2 | 1738 | Missense Mutation | ATC,GTC | I,V 444 | XP_011542466.1 | |
XM_017001045.1 | 1738 | Missense Mutation | ATC,GTC | I,V 444 | XP_016856534.1 |