Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAATTCTAAACTTTCCCCCAAG[C/T]TGCAAGAAATTGTCTACCCACATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD5L - CD5 molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005894.2 | 1660 | UTR 3 | NP_005885.1 | |||
XM_005245602.1 | 1660 | Intron | XP_005245659.1 | |||
XM_017002806.1 | 1660 | Intron | XP_016858295.1 |