Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATAGACCGGGGCGCCGGCGCC[C/G]ACGCGCTCCGAATAGTTGCCCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615015 MIM: 615046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST3H2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST3H2A - histone cluster 3, H2a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033445.2 | Intron | NP_254280.1 |
HIST3H2BB - histone cluster 3, H2bb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175055.2 | Intron | NP_778225.1 |
MIR4666A - microRNA 4666a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |