Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTGCTGCCACAGCTCTGGAGG[C/T]TGCTGCTGACCTGGGCCATGAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612619 | ||||||||||||||||||||
Literature Links: |
CRCT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRCT1 - cysteine rich C-terminal 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE5A - late cornified envelope 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178438.4 | 524 | Silent Mutation | GGC,GGT | G,G 116 | NP_848525.1 |