Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATAAGACCAGCTTCTTACTTGT[C/T]GCCTCTGAGTATAGGGTCTTCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606388 | ||||||||||||||||||||
Literature Links: |
THEM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THEM4 - thioesterase superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053055.4 | 1105 | Silent Mutation | GCA,GCG | A,A 226 | NP_444283.2 | |
XM_011509142.2 | 1105 | Intron | XP_011507444.1 |