Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGACTTCATGGAATAATCAAG[C/T]GGCATGACTACATTAGTTGTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171835 MIM: 612023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PFKFB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PFKFB2 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YOD1 - YOD1 deubiquitinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276320.1 | 4263 | UTR 3 | NP_001263249.1 | |||
NM_018566.3 | 4263 | UTR 3 | NP_061036.3 |