Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGTGAACATTGTAGAAATATA[C/T]TAATGTCTTTGTGAGTACAGATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDR2 - discoidin domain receptor tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014796.1 | Intron | NP_001014796.1 | ||||
NM_006182.2 | Intron | NP_006173.2 | ||||
XM_006711344.3 | Intron | XP_006711407.1 | ||||
XM_011509586.2 | Intron | XP_011507888.1 | ||||
XM_011509587.2 | Intron | XP_011507889.1 | ||||
XM_011509588.2 | Intron | XP_011507890.1 |