Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAATCAACAGGTCCCCAAGGTC[A/G]TAGGAGTCGAAGAGATCGCTGATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600426 | ||||||||||||||||||||
Literature Links: |
E2F2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
E2F2 - E2F transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004091.3 | 1192 | Silent Mutation | TAC,TAT | Y,Y 429 | NP_004082.1 | |
XM_005245748.3 | 1192 | Silent Mutation | TAC,TAT | Y,Y 241 | XP_005245805.1 | |
XM_005245749.3 | 1192 | Silent Mutation | TAC,TAT | Y,Y 233 | XP_005245806.1 | |
XM_011540868.2 | 1192 | Silent Mutation | TAC,TAT | Y,Y 427 | XP_011539170.1 | |
XM_011540870.2 | 1192 | Intron | XP_011539172.1 | |||
XM_011540871.2 | 1192 | Silent Mutation | TAC,TAT | Y,Y 233 | XP_011539173.1 |