Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCGCAGTTCCACGTCAAGTCCG[G/T]CCTGCAGATCAAGAAGAACGCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602006 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPKAPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPKAPK2 - mitogen-activated protein kinase-activated protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004759.4 | 362 | Missense Mutation | GGC,GTC | G,V 51 | NP_004750.1 | |
NM_032960.3 | 362 | Missense Mutation | GGC,GTC | G,V 51 | NP_116584.2 | |
XM_005273353.3 | 362 | Missense Mutation | GGC,GTC | G,V 51 | XP_005273410.1 | |
XM_017002810.1 | 362 | Intron | XP_016858299.1 |