Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATAAGCTGGATTTCCACTGTG[C/T]ATTATCATGTGTCTTCTAACACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf229 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf229 - chromosome 1 open reading frame 229 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF669 - zinc finger protein 669 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142572.1 | 611 | Missense Mutation | ATA,ATG | I,M 157 | NP_001136044.1 | |
NM_024804.2 | 611 | Missense Mutation | ATA,ATG | I,M 243 | NP_079080.2 | |
XM_005273270.2 | 611 | Missense Mutation | ATA,ATG | I,M 120 | XP_005273327.1 | |
XM_005273271.2 | 611 | Missense Mutation | ATA,ATG | I,M 120 | XP_005273328.1 |