Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGGACAGCTGAAAGAAGTAGCA[C/T]TAAGAAAGATCATCCGAGCAGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606920 MIM: 604396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS2 - ceramide synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022075.4 | 2170 | UTR 3 | NP_071358.1 | |||
NM_181746.3 | 2170 | UTR 3 | NP_859530.1 | |||
XM_011509451.2 | 2170 | UTR 3 | XP_011507753.1 | |||
XM_011509452.2 | 2170 | UTR 3 | XP_011507754.1 |
SETDB1 - SET domain bifurcated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145415.1 | 2170 | Intron | NP_001138887.1 | |||
NM_001243491.1 | 2170 | Intron | NP_001230420.1 | |||
NM_012432.3 | 2170 | Intron | NP_036564.3 | |||
XM_005245641.3 | 2170 | Intron | XP_005245698.1 | |||
XM_017002953.1 | 2170 | Intron | XP_016858442.1 | |||
XM_017002954.1 | 2170 | Intron | XP_016858443.1 | |||
XM_017002955.1 | 2170 | Intron | XP_016858444.1 |