Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTGCTCAGCGGGTTGGCCACG[C/T]GGCCCAGGAAGTGCAGGGCAGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 106150 MIM: 606974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGT - angiotensinogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000029.3 | 1938 | Missense Mutation | CAC,CGC | H,R 477 | NP_000020.1 |
COG2 - component of oligomeric golgi complex 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |