Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGTGACACGACACCTCAGTCAC[C/T]GGGCCAACAACTTCAAACGACACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605763 | ||||||||||||||||||||
Literature Links: |
SLC45A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC45A1 - solute carrier family 45 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080397.2 | 250 | Missense Mutation | CGG,TGG | R,W 76 | NP_001073866.2 | |
XM_011541530.1 | 250 | Missense Mutation | CGG,TGG | R,W 76 | XP_011539832.1 | |
XM_011541531.1 | 250 | Missense Mutation | CGG,TGG | R,W 76 | XP_011539833.1 |