Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAGAGGAAGAAATAGAAAAAGG[C/T]GTAGAGATAGAAGAGAAGATAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPCS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPCS - phosphopantothenoylcysteine synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077447.2 | 732 | Silent Mutation | GGC,GGT | G,G 115 | NP_001070915.1 | |
NM_001287506.1 | 732 | Silent Mutation | GGC,GGT | G,G 115 | NP_001274435.1 | |
NM_001287507.1 | 732 | Silent Mutation | GGC,GGT | G,G 82 | NP_001274436.1 | |
NM_001287508.1 | 732 | Silent Mutation | GGC,GGT | G,G 115 | NP_001274437.1 | |
NM_001287509.1 | 732 | Silent Mutation | GGC,GGT | G,G 115 | NP_001274438.1 | |
NM_001287510.1 | 732 | Silent Mutation | GGC,GGT | G,G 115 | NP_001274439.1 | |
NM_001287511.1 | 732 | Intron | NP_001274440.1 | |||
NM_024664.3 | 732 | Silent Mutation | GGC,GGT | G,G 288 | NP_078940.2 |
ZMYND12 - zinc finger MYND-type containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |