Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGCTCCCCGTCAGGGACCCAA[A/G]TGTCCTCACCGTGCTGTGAACAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614989 MIM: 138390 | ||||||||||||||||||||
Literature Links: |
EPS8L3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS8L3 - EPS8 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319952.1 | 1798 | Missense Mutation | CTT,TTT | L,F 488 | NP_001306881.1 | |
NM_024526.3 | 1798 | Missense Mutation | CTT,TTT | L,F 521 | NP_078802.2 | |
NM_133181.3 | 1798 | Missense Mutation | CTT,TTT | L,F 551 | NP_573444.2 | |
NM_139053.2 | 1798 | Missense Mutation | CTT,TTT | L,F 552 | NP_620641.1 | |
XM_011542132.1 | 1798 | Missense Mutation | CTT,TTT | L,F 560 | XP_011540434.1 | |
XM_011542133.1 | 1798 | Missense Mutation | CTT,TTT | L,F 559 | XP_011540435.1 | |
XM_011542134.2 | 1798 | Missense Mutation | CTT,TTT | L,F 526 | XP_011540436.1 | |
XM_011542135.2 | 1798 | Missense Mutation | CTT,TTT | L,F 496 | XP_011540437.1 | |
XM_017002327.1 | 1798 | Intron | XP_016857816.1 | |||
XM_017002328.1 | 1798 | Missense Mutation | CTT,TTT | L,F 530 | XP_016857817.1 | |
XM_017002329.1 | 1798 | Missense Mutation | CTT,TTT | L,F 522 | XP_016857818.1 |
GSTM3 - glutathione S-transferase mu 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |