Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACGGCCTGGATGTTGGGCAGGAC[A/G]CCACCCTGCGCGATGGTCACGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615015 MIM: 615046 | ||||||||||||||||||||
Literature Links: |
HIST3H2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST3H2A - histone cluster 3, H2a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033445.2 | Intron | NP_254280.1 |
HIST3H2BB - histone cluster 3, H2bb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175055.2 | Intron | NP_778225.1 |
MIR4666A - microRNA 4666a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |