Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTCCCGCCAAGAGCACTACGGC[C/T]CCAGGGACACCCTCGTCAAACCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605127 MIM: 601914 | ||||||||||||||||||||
Literature Links: |
OPTC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OPTC - opticin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014359.3 | 407 | Silent Mutation | GCC,GCT | A,A 97 | NP_055174.1 | |
XM_011509406.2 | 407 | Silent Mutation | GCC,GCT | A,A 97 | XP_011507708.1 |
PRELP - proline and arginine rich end leucine rich repeat protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |