Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTGAGAACAACGTGGGCCTTAT[A/C]ACACTGGCTAAGTATGGGGGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601648 | ||||||||||||||||||||
Literature Links: |
PSMD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMD4 - proteasome 26S subunit, non-ATPase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002810.2 | Intron | NP_002801.1 | ||||
XM_005245354.2 | Intron | XP_005245411.1 | ||||
XM_006711464.3 | Intron | XP_006711527.1 | ||||
XM_017001856.1 | Intron | XP_016857345.1 |