Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTCCAGAGCAGCGCCAACTACG[A/C]GGAGAACTTCATCCAGAGTATCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGM1 - phosphoglucomutase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172818.1 | 320 | Intron | NP_001166289.1 | |||
NM_001172819.1 | 320 | Intron | NP_001166290.1 | |||
NM_002633.2 | 320 | Missense Mutation | GAG,GCG | E,A 36 | NP_002624.2 |