Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTCTCAGTCTCTCATATGTT[A/T]CCATGTCAGGTTTGATCTGCTTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607090 | ||||||||||||||||||||
Literature Links: |
SYF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYF2 - SYF2 pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015484.4 | 369 | Missense Mutation | GAA,GTA | E,V 147 | NP_056299.1 | |
NM_207170.3 | 369 | Missense Mutation | GAA,GTA | E,V 105 | NP_997053.1 |