Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAACTGTTGCCCACATGCAGCCC[C/T]GGACACAGCCCCTAGCCCAAACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610595 MIM: 607377 MIM: 607646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLAD1 - flavin adenine dinucleotide synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184891.1 | 32 | Intron | NP_001171820.1 | |||
NM_001184892.1 | 32 | Intron | NP_001171821.1 | |||
NM_025207.4 | 32 | Intron | NP_079483.3 | |||
NM_201398.2 | 32 | Intron | NP_958800.1 |
LENEP - lens epithelial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018655.2 | 32 | Silent Mutation | CGG,TGG | R,W 4 | NP_061125.1 |
ZBTB7B - zinc finger and BTB domain containing 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |