Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTGTATGTTACTATATTTTGTG[C/T]ATTTTCGGTGTTGTAAAATACTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf194 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf194 - chromosome 1 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1324 - KIAA1324 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267048.1 | Intron | NP_001253977.1 | ||||
NM_001284352.1 | Intron | NP_001271281.1 | ||||
NM_001284353.1 | Intron | NP_001271282.1 | ||||
NM_020775.4 | Intron | NP_065826.2 | ||||
XM_011541825.1 | Intron | XP_011540127.1 | ||||
XM_011541826.2 | Intron | XP_011540128.1 | ||||
XM_011541827.1 | Intron | XP_011540129.1 | ||||
XM_017001883.1 | Intron | XP_016857372.1 |