Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTCATGGGCCCTGCCTCTGCT[C/G]CTAGCCTCCCCTGCAGAGTCGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606510 | ||||||||||||||||||||
Literature Links: |
FCRL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCRL3 - Fc receptor like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320333.1 | 2431 | Missense Mutation | AGC,AGG | S,R 708 | NP_001307262.1 | |
NM_052939.3 | 2431 | Missense Mutation | AGC,AGG | S,R 708 | NP_443171.2 | |
XM_006711145.1 | 2431 | Missense Mutation | AGC,AGG | S,R 708 | XP_006711208.1 |