Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTTACCAGGATCAGTTCCCCAT[C/T]GTTGGTCAGTTCTCTGGTCCACGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 180231 | ||||||||||||||||||||
Literature Links: |
CRABP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRABP2 - cellular retinoic acid binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199723.1 | 570 | Missense Mutation | AAT,GAT | N,D 117 | NP_001186652.1 | |
NM_001878.3 | 570 | Missense Mutation | AAT,GAT | N,D 117 | NP_001869.1 | |
XM_017000343.1 | 570 | Missense Mutation | AAT,GAT | N,D 117 | XP_016855832.1 |