Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGACGTCAGCGCCTCCTCGTCG[A/C]TGAGTTCGGCGTCCATGCCGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CITED4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CITED4 - Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133467.2 | 629 | Missense Mutation | AGC,ATC | S,I 137 | NP_597724.1 |