Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGTCTGGGTGTGGAGAAGCGC[A/G]TGAGCCAGCCTTTGCCCCGTGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612801 MIM: 609275 | ||||||||||||||||||||
Literature Links: |
IARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IARS2 - isoleucyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3GAP2 - RAB3 GTPase activating non-catalytic protein subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012414.3 | 4106 | Silent Mutation | CAC,CAT | H,H 1314 | NP_036546.2 |