Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGGGTCTCTGCCTCAAACTG[C/T]TCACATATTTTGCACATGACGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601924 MIM: 600279 | ||||||||||||||||||||
Literature Links: |
COPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPA - coatomer protein complex subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX19 - peroxisomal biogenesis factor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193644.1 | 717 | Silent Mutation | GAA,GAG | E,E 230 | NP_001180573.1 | |
NM_002857.3 | 717 | Silent Mutation | GAA,GAG | E,E 230 | NP_002848.1 |