Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGGTCCTTGTGCAGTTCATGA[C/T]GGTGTGGCAGCGGTATAGAGAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 185470 | ||||||||||||||||||||
Literature Links: |
SDHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDHB - succinate dehydrogenase complex iron sulfur subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003000.2 | 887 | Missense Mutation | ATC,GTC | I,V 246 | NP_002991.2 |