Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAACCGTGGGTCTCCCCGACCAG[G/T]CCGTCCCCACCGGCTGCTGGTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610378 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIS1 - GLIS family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147193.2 | 2374 | Silent Mutation | NP_671726.2 | |||
XM_017000408.1 | 2374 | Silent Mutation | XP_016855897.1 | |||
XM_017000409.1 | 2374 | Silent Mutation | XP_016855898.1 | |||
XM_017000410.1 | 2374 | Silent Mutation | XP_016855899.1 | |||
XM_017000411.1 | 2374 | Intron | XP_016855900.1 | |||
XM_017000412.1 | 2374 | Intron | XP_016855901.1 |