Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCCGTGCTCGGAGAGGACCCT[A/G]TACTAAAGCCACCACCATCATTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMIM12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMIM12 - small integral membrane protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164824.1 | 4382 | UTR 3 | NP_001158296.1 | |||
NM_001164825.1 | 4382 | UTR 3 | NP_001158297.1 | |||
NM_001320261.1 | 4382 | UTR 3 | NP_001307190.1 | |||
NM_138428.5 | 4382 | UTR 3 | NP_612437.3 | |||
XM_005270403.4 | 4382 | Intron | XP_005270460.1 | |||
XM_011540553.2 | 4382 | Intron | XP_011538855.1 | |||
XM_011540554.2 | 4382 | Intron | XP_011538856.2 | |||
XM_011540555.2 | 4382 | Intron | XP_011538857.1 | |||
XM_017000166.1 | 4382 | Intron | XP_016855655.1 |