Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTTGGCTGGCTCATCAATGAC[C/G]AGCTTCCCTTCCTCGTCGCTGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600339 MIM: 611836 | ||||||||||||||||||||
Literature Links: |
HDGF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HDGF - hepatoma-derived growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126050.1 | 525 | Silent Mutation | CTC,CTG | L,L 155 | NP_001119522.1 | |
NM_001126051.1 | 525 | Silent Mutation | CTC,CTG | L,L 132 | NP_001119523.1 | |
NM_001319186.1 | 525 | Silent Mutation | CTC,CTG | L,L 162 | NP_001306115.1 | |
NM_001319187.1 | 525 | Silent Mutation | CTC,CTG | L,L 107 | NP_001306116.1 | |
NM_001319188.1 | 525 | Silent Mutation | CTC,CTG | L,L 107 | NP_001306117.1 | |
NM_004494.2 | 525 | Silent Mutation | CTC,CTG | L,L 139 | NP_004485.1 | |
XM_011509453.2 | 525 | Silent Mutation | CTC,CTG | L,L 107 | XP_011507755.2 | |
XM_011509454.2 | 525 | Silent Mutation | CTC,CTG | L,L 107 | XP_011507756.1 | |
XM_011509455.2 | 525 | Silent Mutation | CTC,CTG | L,L 107 | XP_011507757.1 |
MRPL24 - mitochondrial ribosomal protein L24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |