Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGGGTAGTCACGGAGGTGGCT[A/G]ACGTGTGCAGATGACACGAAATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF220 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF220 - ring finger protein 220 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM53 - transmembrane protein 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300746.1 | 1105 | Silent Mutation | GTC,GTT | V,V 180 | NP_001287675.1 | |
NM_001300747.1 | 1105 | Silent Mutation | GTC,GTT | V,V 223 | NP_001287676.1 | |
NM_001300748.1 | 1105 | Silent Mutation | GTC,GTT | V,V 222 | NP_001287677.1 | |
NM_024587.3 | 1105 | Silent Mutation | GTC,GTT | V,V 253 | NP_078863.2 | |
XM_011542138.2 | 1105 | Silent Mutation | GTC,GTT | V,V 136 | XP_011540440.1 |