Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTCATTTGATAAGTTTAACAA[A/G]TTATCCAGTGGCGTTACATTATTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf141 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf141 - chromosome 1 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276351.1 | 1397 | Silent Mutation | AAC,AAT | N,N 388 | NP_001263280.1 | |
NM_001276352.1 | 1397 | UTR 3 | NP_001263281.1 | |||
XM_011541465.2 | 1397 | Silent Mutation | AAC,AAT | N,N 459 | XP_011539767.1 | |
XM_011541466.2 | 1397 | Silent Mutation | AAC,AAT | N,N 459 | XP_011539768.1 | |
XM_011541467.1 | 1397 | Silent Mutation | AAC,AAT | N,N 446 | XP_011539769.1 | |
XM_011541469.1 | 1397 | Silent Mutation | AAC,AAT | N,N 321 | XP_011539771.1 | |
XM_011541472.1 | 1397 | Intron | XP_011539774.1 | |||
XM_011541473.2 | 1397 | Intron | XP_011539775.1 | |||
XM_011541475.1 | 1397 | Intron | XP_011539777.1 | |||
XM_017001276.1 | 1397 | Silent Mutation | AAC,AAT | N,N 459 | XP_016856765.1 | |
XM_017001277.1 | 1397 | Intron | XP_016856766.1 |