Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCTGCCCATAAAGGACCTGAT[C/T]GCTGGTGCCAGACTGGGATTGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138971 MIM: 611979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF3R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF3R - colony stimulating factor 3 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000760.3 | 2826 | Missense Mutation | AAT,GAT | N,D 748 | NP_000751.1 | |
NM_156039.3 | 2826 | Missense Mutation | AAT,GAT | N,D 775 | NP_724781.1 | |
NM_172313.2 | 2826 | Missense Mutation | AAT,GAT | N,D 748 | NP_758519.1 | |
XM_005270493.1 | 2826 | Missense Mutation | AAT,GAT | N,D 747 | XP_005270550.1 | |
XM_011540748.2 | 2826 | Missense Mutation | AAT,GAT | N,D 775 | XP_011539050.1 | |
XM_011540749.1 | 2826 | Missense Mutation | AAT,GAT | N,D 774 | XP_011539051.1 | |
XM_011540750.1 | 2826 | Missense Mutation | AAT,GAT | N,D 551 | XP_011539052.1 | |
XM_017000370.1 | 2826 | Missense Mutation | AAT,GAT | N,D 775 | XP_016855859.1 |
MRPS15 - mitochondrial ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |