Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGTCATGGCGACCGAGCAGAGGC[C/T]TTTCCACCTGGTGGTGTTCGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SCCPDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCCPDH - saccharopine dehydrogenase (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016002.2 | 367 | Missense Mutation | CCT,CTT | P,L 7 | NP_057086.2 |