Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTGTTAAAGGAGTGAATACTC[A/C]TAAGAACCCCTGTAAGGTATGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCTF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHCTF1 - AT-hook containing transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323342.1 | Intron | NP_001310271.1 | ||||
NM_001323343.1 | Intron | NP_001310272.1 | ||||
NM_015446.4 | Intron | NP_056261.4 | ||||
XM_006711758.1 | Intron | XP_006711821.1 | ||||
XM_011544156.2 | Intron | XP_011542458.2 | ||||
XM_011544157.2 | Intron | XP_011542459.2 |