Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCAGGATCTGCTCTTGAATCCG[G/T]TTTTACAAACTCTTGAGATCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EYA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EYA3 - EYA transcriptional coactivator and phosphatase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPDL3B - sphingomyelin phosphodiesterase acid like 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XKR8 - XK related 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018053.3 | Intron | NP_060523.2 | ||||
XM_011541679.2 | Intron | XP_011539981.1 | ||||
XM_011541680.2 | Intron | XP_011539982.1 |