Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTAGAATTATGGCGACCTCCG[C/T]GACGTCGCCGCACGCGCCTGGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609321 | ||||||||||||||||||||
Literature Links: |
SASS6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SASS6 - SAS-6 centriolar assembly protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304829.1 | 103 | Intron | NP_001291758.1 | |||
NM_194292.2 | 103 | Intron | NP_919268.1 | |||
XM_017000486.1 | 103 | Intron | XP_016855975.1 |
TRMT13 - tRNA methyltransferase 13 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019083.2 | 103 | Missense Mutation | GCG,GTG | A,V 5 | NP_061956.2 | |
XM_005270945.3 | 103 | Missense Mutation | GCG,GTG | A,V 5 | XP_005271002.1 | |
XM_005270946.2 | 103 | UTR 5 | XP_005271003.1 | |||
XM_017001504.1 | 103 | Missense Mutation | GCG,GTG | A,V 5 | XP_016856993.1 | |
XM_017001505.1 | 103 | Missense Mutation | GCG,GTG | A,V 5 | XP_016856994.1 |